Order Entry
France
ContactUsLinkComponent
Sodium iodide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sodium iodide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0293.5000
Supplier:  Avantor
CAS Number:  
Sodium iodide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0293.5000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay (dried basis):
    min. 99.0 %
  • Barium (Ba):
    max. 0.002 %
  • Iron (Fe):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Insoluble Matter:
    max. 0.005 %
  • Iodate (IO3):
    max. 4 ppm
  • Nitrogen Compounds (as N):
    max. 0.001 %
  • pH of 5% Solution at 25°C:
    7.5 - 9.5
  • Phosphate (PO4):
    max. 0.002 %
  • Sulfate (SO4):
    max. 150 ppm

Specification Test Results

About this item

 :
 :149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
 :GHS09
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532