
Cesium chloride 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1290 °C (760 mmHg)
Melting Pt:646 °C
Density:3,7808 g/cm³ (20 °C)
EINECS:231-600-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystals
- Titration Argentometric:≥99.0 %
- Rhodium (Rh):≥24.0 %
Specification Test Results
Frequently Bought Together