Order Entry
France
ContactUsLinkComponent
 
Irritant
4,5-Difluorophthalic anhydride, Sigma-Aldrich®
Catalog # 8.14389.0001
Supplier:  Merck
CAS Number:  
Irritant
4,5-Difluorophthalic anhydride, Sigma-Aldrich®
Catalog # 8.14389.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    92 - 96 °C
  • Molar Mass:
    184.09 g/mol
  • Boiling Point:
    92 - 96 °C (14 hPa)
  • Flash Point:
    73 °C
  • Density:
    0.975 g/cm3
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :184.1 g/mol
UN:3261
 :GHS07
Storage Temperature:Ambient
CAS Number:18959-30-3
Formula:C₈H₂F₂O₃
MDL Number:MFCD08695033