TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Millipore®
Supplier: Merck
Certificates
About this item
Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid
CAS Number:29915-38-6
:
Formula:C₇H₁₇NO₆S
MDL Number:MFCD00007538
Molecular Weight:243.28 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:249-954-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together