4-Nitrophthalic anhydride tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5466-84-2
:
Formula:C₈H₃NO₅
MDL Number:MFCD00005922
:193.12 g/mol
Melting Pt:116…121 °C
EINECS:226-776-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together