Order Entry
France
Orders LinkContactUsLinkComponent
 

4-Phenylbenzaldehyde

Glass bottle for solids, 25 g
Catalog # MOLEM60707293
Irritant
Marketsource
Supplier:   Molekula
4-Phenylbenzaldehyde
Catalog # MOLEM60707293
Supplier:  Molekula
Marketsource
CAS Number:  3218-36-8
4-Phenylbenzaldehyde

Glass bottle for solids, 25 g

CAS Number:  3218-36-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
  • CAS Number:3218-36-8
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00006947
  • Molecular Weight:182.22 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:221-742-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications