Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Di-tert-butylphenol
2,6-Di-tert-butylphenol
2,6-Di-tert-butylphenol
EHERC12254700
:
EHRENSTORFER
:
2,6-Di-tert-butylphenol
EHERC12254700
:
EHRENSTORFER
:
Pk:
0,1 g
Seq:
3020
:
:
251 °C (1013 hPa)
:
34…37 °C
:
0.914 g/cm³ (20 °C)
:
107 °C
:
Ambient
:
204-884-0
:
3077
:
9,III
:
206.33 g/mol
:
GHS09
:
128-39-2
:
C₁₄H₂₂O
:
MFCD00008820