Order Entry
France
ContactUsLinkComponent
5-Nitro-m-xylene
5-Nitro-m-xylene
Catalog # MOLEM19086651
Supplier:  Molekula
CAS Number:  
5-Nitro-m-xylene
Catalog # MOLEM19086651
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:251 °C (1013 hPa)
Melting Pt:70 °C
EINECS:202-732-8
UN:3447
ADR:6.1,II
CAS Number:99-12-7
Formula:C₈H₉NO₂
MDL Number:MFCD00007269