Order Entry
France
Orders LinkContactUsLinkComponent
α-Chloro-m-xylene 98%

α-Chloro-m-xylene 98%

Irritant
Corrosive
α-Chloro-m-xylene 98%
Certificates
α-Chloro-m-xylene 98%

2 Options Available Below

Product Details & Documents
Synonyms:  1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride
  • CAS Number:620-19-9
  • Formula:C₈H₉Cl
  • MDL Number:MFCD00000909
  • Molecular Weight:140.61 g/mol
  • Merck Index:12,10224
  • Boiling Pt:196 °C (1013 hPa)
  • Density:1.064 g/cm³
  • Flash Pt:75 °C
  • Storage Temperature:Refrigerator
  • EINECS:210-628-9
  • UN:3265
  • ADR:8,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5335 to 1.5365 (20°C, 589 nm)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 60,30 €
    60,30 €
    1 * 25 mL
       60,30 €

  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 116,00 €
    116,00 €
    1 * 100 mL
       116,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.