
Hydrochloric acid 0.05 M (0.05 N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.0 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1789
ADR:8,III
:36.46 g/mol
:GHS05
Specification Test Results
- Vapor Pressure:190 mmHg ( 25 °C)
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together