Order Entry
France
ContactUsLinkComponent
1,1-Dichloroethylene stabilised, Msynth® plus
Flammable
Health
Irritant
1,1-Dichloroethylene stabilised, Msynth® plus
Catalog # 8.45033.9999
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
1,1-Dichloroethylene stabilised, Msynth® plus
Catalog # 8.45033.9999
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Solubility:
    2.5 g/l (25 °C)
  • Melting Point:
    -122 °C
  • Molar Mass:
    96.94 g/mol
  • Boiling Point:
    31.2 - 31.7 °C (1013 hPa)
  • Vapor Pressure:
    665 hPa (20 °C)
  • Flash Point:
    -25 °C
  • Refractive Index:
    1.4254 (20 °C, 589 nm)
  • Explosion Limit:
    8.4 - 16.5 %(V)
  • Density:
    1.21 g/cm3 (20 °C)
  • Ignition Point:
    520 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
Merck Index:12,10134
Boiling Pt:31.7 °C (1013 hPa)
Melting Pt:–122 °C
Density:1.21 g/cm³ (20 °C)
Flash Pt:–25 °C
ADR:3,I
 :M-Clarity™ quality level = MQ400
 :Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
 :96.94 g/mol
Storage Temperature:Refrigerator
EINECS:200-864-0
UN:1303
 :GHS02
 :GHS08
 :GHS07
CAS Number:75-35-4
Formula:C₂H₂Cl₂
MDL Number:MFCD00011653