Order Entry
France
ContactUsLinkComponent
 
 
 
Mesityl oxide, Sigma-Aldrich®
 :  Merck
 
 
 
Mesityl oxide, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    30 g/l (20 °C)
  • Melting Point:
    -59 °C
  • Molar Mass:
    98.14 g/mol
  • Boiling Point:
    130 °C
  • Vapor Pressure:
    10.5 hPa (20 °C)
  • Flash Point:
    28 °C
  • Refractive Index:
    1.4445 (20 °C, 589 nm)
  • Explosion Limit:
    1.4 - 10.1 %(V)
  • Density:
    0.85 g/cm3 (20 °C)
  • Ignition Point:
    344 °C

 

 

Frequently Bought Together