

4-Nitroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-01-6
:
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007858
:138.13 g/mol
Merck Index:14,6584
Boiling Pt:260 °C (100 mmHg)
Melting Pt:147…149 °C
Density:1.437
Flash Pt:165 °C (329 °F)
Storage Temperature:Ambient
EINECS:202-810-1
UN:1661
ADR:6.1,II
:GHS06
:GHS08
Related Information
Frequently Bought Together