


N,N-Diethylaniline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91-66-7
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00009042
:149.24 g/mol
Merck Index:13,03140
Boiling Pt:215.5 °C (1013 hPa)
Melting Pt:–38 °C
Density:0.933 g/cm³
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:202-088-8
UN:2432
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5410 to 1.5430 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together