Order Entry
France
ContactUsLinkComponent
2,4-Difluorophenol
2,4-Difluorophenol
Catalog # MOLEM52407368
Supplier:  Molekula
CAS Number:  
2,4-Difluorophenol
Catalog # MOLEM52407368
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :130.09 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:20 °C
Density:1.36 g/cm³ (20 °C)
EINECS:206-688-0
UN:2921
ADR:8,II
CAS Number:367-27-1
Formula:C₆H₄F₂O
MDL Number:MFCD00009715