

1,2,4-Trichlorobenzene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-82-1
:
Formula:C₆H₃Cl₃
MDL Number:MFCD00000547
:181.45 g/mol
Merck Index:13,09704
Boiling Pt:214.4 °C (1013 hPa)
Melting Pt:17 °C
Density:1.45 g/cm³ (20 °C)
EINECS:204-428-0
UN:2321
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:99 °C
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.049 g/l (20 °C)
- Melting Point:17 °C
- Molar Mass:181.45 g/mol
- Boiling Point:213.5 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (20 °C)
- Flash Point:110 °C
- Explosion Limit:2.5 - 6.6 %(V)
- Density:1.45 g/cm3 (20 °C)
- Ignition Point:571 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together