Order Entry
France
ContactUsLinkComponent
Salicylic acid ≥99%, GPR RECTAPUR®
Salicylic acid ≥99%, GPR RECTAPUR®
Catalog # 20655.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Salicylic acid ≥99%, GPR RECTAPUR®
Catalog # 20655.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Melting point:
    158 - 161 °C
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :138.12 g/mol
Merck Index:13,08411
Boiling Pt:336 °C (1013 hPa)
Melting Pt:157…159 °C
Density:1.443 g/cm³ (20 °C)
Flash Pt:157 °C
EINECS:200-712-3
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:69-72-7
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439