Order Entry
France
ContactUsLinkComponent
Salicylic acid
Salicylic acid
Catalog # MOLEM34871437
Supplier:  Molekula
CAS Number:  
undefined
Salicylic acid
Catalog # MOLEM34871437
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :138.12 g/mol
Merck Index:13,08411
Boiling Pt:336 °C (1013 hPa)
Melting Pt:157…159 °C
Density:1.443 g/cm³ (20 °C)
Flash Pt:157 °C
EINECS:200-712-3
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:69-72-7
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439