Order Entry
France
ContactUsLinkComponent
Salicylic acid, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Salicylic acid, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Catalog # 1.00635.9019
CAS Number:  
undefined
Salicylic acid, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Catalog # 1.00635.9019
CAS Number:  

Specifications

  • Pk:
    15 kg
  • Seq:
    4211

Specifications

About this item

 :
 :138.12 g/mol
Merck Index:13,08411
Boiling Pt:336 °C (1013 hPa)
Melting Pt:157…159 °C
Density:1.443 g/cm³ (20 °C)
Flash Pt:157 °C
EINECS:200-712-3
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:69-72-7
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439