Salicylic acid
Catalog #
Product Details & Documents
Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
- CAS Number:69-72-7
- Formula:(HO)C₆H₄CO₂H
- MDL Number:MFCD00002439
- Molecular Weight:138.12 g/mol
- Merck Index:13,08411
- Boiling Pt:336 °C (1013 hPa)
- Melting Pt:157…159 °C
- Density:1.443 g/cm³ (20 °C)
- Flash Pt:157 °C
- Storage Temperature:Ambient
- EINECS:200-712-3



