Order Entry
France
ContactUsLinkComponent
2,3,4-Trimethoxybenzaldehyde 99%
2,3,4-Trimethoxybenzaldehyde 99%
Certificates
undefined
2,3,4-Trimethoxybenzaldehyde 99%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    ≥36.5 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together