Order Entry
France
ContactUsLinkComponent
6-Methoxysalicylaldehyde
6-Methoxysalicylaldehyde
Catalog # MOLE86881166-1G
Supplier:  Molekula
CAS Number:  
6-Methoxysalicylaldehyde
Catalog # MOLE86881166-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.15 g/mol
Storage Temperature:Ambient
EINECS:211-844-6
CAS Number:700-44-7
Formula:C₈H₈O₃
MDL Number:MFCD00151830