Order Entry
France
ContactUsLinkComponent
Cholecalciferol (Vitamin D3)
Cholecalciferol (Vitamin D3)
Catalog # MOLE28904887-5G
Supplier:  Molekula
CAS Number:  
Cholecalciferol (Vitamin D3)
Catalog # MOLE28904887-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :384.65 g/mol
EINECS:200-673-2
UN:2811
ADR:6.1,II
CAS Number:67-97-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131