Order Entry
France
ContactUsLinkComponent
Organic reference standard, m-Xylene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, m-Xylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17945100ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, m-Xylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17945100ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    108-38-3
  • Description:
    m-Xylene 100 µg/ml in methanol
  • Formula:
    C₈H₁₀
  • Description:
    m-Xylene

Specifications

About this item