Organic reference standard, (S)-(-)-Nicotine 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15520000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:54-11-5
- Description:(S)-(-)-Nicotine 100 µg/ml in methanol
- Formula:C₁₀H₁₄N₂
- Description:(S)-(-)-Nicotine