Order Entry
France
Orders LinkContactUsLinkComponent
 

Ethyl tricosanoate

Supplier:   Cayman Chemical
Ethyl tricosanoate
Supplier:  Cayman Chemical
Certificates
Ethyl tricosanoate

4 Options Available Below

Product Details & Documents
Synonyms:  Tricosanoic acid ethyl ester
  • CAS Number:18281-07-7
  • Formula:C₂₅H₅₀O₂
  • Molecular Weight:382.67 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 27,40 €
    27,40 €
    1 * 5 mg
       27,40 €

  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 47,60 €
    47,60 €
    1 * 10 mg
       47,60 €

  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 184,00 €
    184,00 €
    1 * 50 mg
       184,00 €

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 316,00 €
    316,00 €
    1 * 100 mg
       316,00 €