Order Entry
France
ContactUsLinkComponent
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Health
Environment
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Environment
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Boiling point:
    120 - 122 °C
  • Density (20/4):
    1.620 - 1.630
  • Free acidity:
    Max. 0.0003 meq/g
  • Refractive index (20°C):
    1.503 - 1.505
  • Evaporation residue:
    Max. 10 ppm
  • Free chlorine:
    Max. 2 ppm
  • Water:
    Max. 50 ppm

Specification Test Results

Frequently Bought Together