

Tetrachloroethylene 99+%, Ultrapure for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-18-4
:
Formula:C₂Cl₄
MDL Number:MFCD00000834
:165.83 g/mol
Merck Index:14,9190
Boiling Pt:120…121 °C
Melting Pt:–22 °C
Density:1.623
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
:GHS08
:GHS09
Frequently Bought Together