Benzyl acetate for synthesis, Sigma-Aldrich®
: Merck
:140-11-4
:
:C₉H₁₀O₂
:MFCD00008712
:150.18 g/mol
:13,01125
:206 °C (1013 hPa)
:–51 °C
:1.04 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:95 °C
:Ambient
:205-399-7
- Solubility:(20 °C) slightly soluble
- Melting Point:-51 °C
- Molar Mass:150.17 g/mol
- Boiling Point:205 - 207 °C
- Vapor Pressure:1 hPa (45 °C)
- Flash Point:95 °C
- Refractive Index:1.5006 (20 °C, 589 nm)
- Explosion Limit:0.9 - 8.4 %(V)
- Density:1.06 g/cm3 (20 °C)
- Ignition Point:460 °C
- Synonyms:Acetic acid benzyl ester
- Hill Formula:C9H10O2
- Chemical Formula:CH3COOCH2C6H5
Frequently Bought Together