Order Entry
France
ContactUsLinkComponent
2,2-Dimethylpentane
  EHERC12728200
 :  
2,2-Dimethylpentane
  EHERC12728200
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
 :
 :100.2 g/mol
 :79 °C (1013 hPa)
 :–123.7 °C
 :0.674 g/cm³ (20 °C)
 :Ambient
 :209-680-5
 :1206
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :590-35-2
 :C₇H₁₆
 :MFCD00009404