3-Methylbutanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:563-80-4
:
Formula:C₅H₁₀O
MDL Number:MFCD00008919
Merck Index:12,06164
Boiling Pt:94 °C (1013 hPa)
Melting Pt:–92 °C
Density:0.8046 g/cm³ (20 °C)
Flash Pt:–3 °C
Storage Temperature:Refrigerator
EINECS:209-264-3
UN:2397
ADR:3,II
:M-Clarity™ quality level = MQ200
:86.13 g/mol
:GHS02
Specification Test Results
- Solubility:6 g/l (20 °C)
- Melting Point:-92 °C
- Molar Mass:86.13 g/mol
- Boiling Point:93 - 95 °C (1013 hPa)
- Vapor Pressure:53.33 hPa (19.9 °C)
- Flash Point:-1 °C
- Refractive Index:1.388 (20 °C, 589 nm)
- Explosion Limit:1.5 - 9 %(V)
- Density:0.80 g/cm3 (20 °C)
- Ignition Point:475 °C
- Evaporation number:5.6
Specification Test Results
Frequently Bought Together