Order Entry
France
ContactUsLinkComponent
n-Pentane ≥99.0% (by GC), CHROMASOLV™ for residue analysis (of high-volatile halogenated hydrocarbons), Riedel-de Haën™
Irritant
Health
Flammable
Environment
n-Pentane ≥99.0% (by GC), CHROMASOLV™ for residue analysis (of high-volatile halogenated hydrocarbons), Riedel-de Haën™
Catalog # 76866-1L
CAS Number:  
undefined
Irritant
Health
Flammable
Environment
n-Pentane ≥99.0% (by GC), CHROMASOLV™ for residue analysis (of high-volatile halogenated hydrocarbons), Riedel-de Haën™
Catalog # 76866-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Explosion Limit:
    ~8.3 %
  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for halogen hydrocarbons:
    Max. 5 µg/l
  • Refractive Index:
    n20/D 1.358 (lit.)
  • Autoignition Temperature:
    500 °F

Specification Test Results

About this item

 :
 :72.15 g/mol
Merck Index:13,07193
Boiling Pt:36-37 °C
Boiling Pt:36…37 °C
Melting Pt:-131 °C
Melting Pt:–131 °C
Density:0.631 g/cm³ (15 °C)
Flash Pt:<-35 °C
Flash Pt:<–35 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
 :GHS07
 :GHS08
 :GHS02
 :GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498