



n-Octane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-65-9
:
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556
:114.23 g/mol
Merck Index:14,6749
Boiling Pt:125…126 °C
Melting Pt:–57 °C
Density:0.703
Flash Pt:15 °C (59 °F)
EINECS:203-892-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together