Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Molecular Weight:32.04 g/mol
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1) II
Merck Index:13,05984
:Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99.9 %
- Acidity:Max. 0.00050 meq/g
- Residue on evaporation:Max. 0.0003 %
- Water (K.F.):Max. 0.0020 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
83679_METHANOL ANHYDROUS (20 PPM WATER)_BSE_REV0
BSE/TSE Certificates
spec_VWRC83679.230 spec_VWRC83679.260 spec_VWRC83679.290
Product specification
VWR® Anhydrous Solvents
Product Literature
Frequently Bought Together