Order Entry
France
Orders LinkContactUsLinkComponent
Methanol, BAKER ANALYZED® ACS, J.T.Baker®

Methanol, BAKER ANALYZED® ACS, J.T.Baker®

Glass bottle, 2,5 L
Catalog # 8046.2500
Flammable
Toxic
Health
   Safer Human and Environmental Health
Supplier:   Avantor
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8046.2500
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  67-56-1
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Methanol, BAKER ANALYZED® ACS, J.T.Baker®

Glass bottle, 2,5 L

CAS Number:  67-56-1
Price
175,46 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44

Documents

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99,8 %
  • Appearance:
    clear
  • Carbonyl Compounds (max. 0.001% each of Acetone, Formaldehyde and Acetaldehyde):
    passes test
  • Color (APHA):
    max. 10
  • Residue after Evaporation (in ppm):
    max. 5
  • Solubility in Water:
    passes test
  • Substances Darkened by H2SO4:
    passes test
  • Substances Reducing KMnO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0,0003
  • Titrable Base (meq/g):
    max. 0,0002
  • Water (H2O):
    max. 0,10 %
  • Ultraviolet Absorbance (1.00-cm path vs water)::
    -
  • at 205 nm:
    max. 1,00
  • at 210 nm:
    max. 0,80
  • at 220 nm:
    max. 0,40
  • at 230 nm:
    max. 0,20
  • at 240 nm :
    max. 0,10
  • at 260 nm:
    max. 0,04
  • at 280-400 nm:
    max. 0,01

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    8046.2500
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.