Order Entry
France
Orders LinkContactUsLinkComponent
 

Solution 192

Flammable
Irritant
Supplier:   PROLABO CMP
Solution 192
Supplier:  PROLABO CMP
Certificates

2 Options Available Below

Product Details & Documents

Hydroxylamine sulphate 2 g + eriochrome black 0,5 g + Isopropanol 300 ml + water QSP 1 000 ml

  • Melting Pt:-89 °C
  • UN:1993
  • ADR:3,II
Product Family Options

Product Information