Order Entry
France
Orders LinkContactUsLinkComponent
(±)-2-Nonanol

(±)-2-Nonanol

Glass bottle for solids, 100 g
Catalog # MOLE25104481-100G
Marketsource
Supplier:   Molekula
(±)-2-Nonanol
Catalog # MOLE25104481-100G
Supplier:  Molekula
Marketsource
CAS Number:  628-99-9
(±)-2-Nonanol

Glass bottle for solids, 100 g

CAS Number:  628-99-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol
  • CAS Number:628-99-9
  • Formula:C₉H₂₀O
  • MDL Number:MFCD00004593
  • Molecular Weight:144.26 g/mol
  • Boiling Pt:193 °C (1013 hPa)
  • Melting Pt:–35 °C
  • Density:0.831 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:211-065-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.