Order Entry
France
ContactUsLinkComponent
1-Octanol for synthesis, Sigma-Aldrich®
Irritant
1-Octanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
1-Octanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.3 g/l (20 °C)
  • Melting Point:
    -16 °C
  • Molar Mass:
    130.23 g/mol
  • Boiling Point:
    195 °C (1013 hPa)
  • Vapor Pressure:
    0.031 hPa (20 °C)
  • Flash Point:
    90 °C
  • Explosion Limit:
    0.8 %(V)
  • Density:
    0.83 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    270 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together