Order Entry
France
ContactUsLinkComponent
1-Hexadecanol for synthesis, Sigma-Aldrich®
1-Hexadecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Hexadecanol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
     - 0.0001 g/l (25 °C)
  • Melting Point:
    46 - 52 °C
  • Molar Mass:
    242.44 g/mol
  • Bulk Density:
    410 - 450 kg/m3
  • Boiling Point:
    305 - 330 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    155 °C DIN 51758
  • Density:
    0.82 g/cm3 (50 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    235 °C DIN 51794

Specification Test Results

Related Information

Frequently Bought Together