

Diethyl ether, dehydrated (max. 0.05% H₂O) ≥99.7%, GPR RECTAPUR® for synthesis
Catalog # 23812.295
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99.7 %
- Assay (Ionol/BHT):4 to 7 ppm
- Evaporation residue:Max. 10 ppm
- Peroxides (as H2O2):Max. 20 ppm
- Water:Max. 0.05 %
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
:GHS02
:GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646