Heptylamine
Catalog # EHERC14135000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:115.22 g/mol
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–18 °C
Density:0,7754 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-895-8
UN:2734
ADR:8,II
CAS Number:111-68-2
Formula:H₃C(CH₂)₅CH₂NH₂
MDL Number:MFCD00008244