Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with ethanol 0.4-1 %
EINECS:200-663-8
REACH:01-2119486657-20
:GHS08
:GHS06
Specification Test Results
- Appearance:Clear colourless liquid
- Assay:99.0 - 99.4 %
- Assay (+ stabilizing agent):Min. 99.8 %
- Insolubility in alcohol:Passes test
- IR Spectrum:Passes test
- Suited for determination with dithizone:Passes test
- Acidity:Max. 0.00014 meq/g
- Boiling point:59.5 - 60.5 °C
- Colouration:Max. 10 APHA
- Density (20/4):1.476 - 1.485
- Density (20/20):1.475-1.481
- Substances coloured by H2SO4:Max. 35 APHA
- Carbon tetrachloride:Max. 100 ppm
- Carbonyl compounds (as CO):Max. 50 ppm
- Dichloromethane:Max. 100 ppm
- Ethanol:0.4 - 1.0 %
- Evaporation residue:Max. 5 ppm
- Free chlorine:Max. 0.3 ppm
- Ionised chlorine:Max. 0.5 ppm
- Tetrachloroethylene:Max. 100 ppm
- Trichloroethylene:Max. 100 ppm
- Water:Max. 100 ppm
- Cl (Chloride):Max. 0.2 ppm
- Al (Aluminium):Max. 0.05 ppm
- B (Boron):Max. 0.01 ppm
- Ba (Barium):Max. 0.01 ppm
- Ca (Calcium):Max. 0.1 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.2 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.01 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.05 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.05 ppm
- Conforms to Reag. Ph.Eur.:Passes test
- Conforms to ACS:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 500 ml Glass bottle for liquids DIN 45 (ZPACE8009063)
Packaging Information
22711_CHLOROFORM ANALAR NORMAPUR ACS_BSE_REV0
BSE/TSE Certificates
spec_VWRC22711.244 spec_VWRC22711.244 spec_VWRC22711.260 spec_VWRC22711.260 spec_VWRC22711.290
Product specification
More
Frequently Bought Together