Chloroform, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:200-663-8
REACH:05-2114614906-45
:GHS06
:GHS08
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:8 g/L (20 °C)
- Melting Point:-63 °C
- Molar Mass:119.38 g/mol
- Boiling Point:61 °C (1013 hPa)
- Vapor Pressure:211 hPa (20 °C)
- Refractive Index:1.45
- Density:1.48 g/cm³ (20 °C)
- pH:(H₂O) no data available
- Ignition Point:982 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Related Information
msds_102447
SDS
ProductPage_102447 ProductPage_102447
Product specification
Solvents for spectroscopy Uvasol
Product Literature
Frequently Bought Together