Chloroform, J.T.Baker®
Catalog # 7152.9200
Supplier: Avantor
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
Specifications
About this item
:
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
:GHS06
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826