Chloroform ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:14,2141
Boiling Pt:60.5…61.5 °C
Melting Pt:–63 °C
Density:1.492
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
:GHS06
:GHS08
:GHS07
Frequently Bought Together