Order Entry
France
ContactUsLinkComponent
1-Chlorobutane ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Flammable
1-Chlorobutane ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83631.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
1-Chlorobutane ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83631.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.70 %
  • Water:
    Max. 0.0100 %
  • Water (KF):
    Max. 0.0005 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Transmittance (227 nm):
    Min. 50.0 %
  • Transmittance (232 nm):
    Min. 80.0 %
  • Transmittance (250 nm):
    Min. 98.0 %
  • Conforms to BDH 15294:
    Passes test

Specification Test Results

About this item

CAS Number:109-69-3
 :
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009
 :92.57 g/mol
Merck Index:13,01559
Boiling Pt:78.4 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.8862 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-696-6
UN:1127
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–17 °C
 :GHS02