Order Entry
France
ContactUsLinkComponent
Acetic acid glacial 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Acetic acid glacial 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Acetic acid glacial 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.8 - 100.5 %
  • Appearance:
    Passes test Ph.Eur.
  • Dilution test:
    Passes test ACS
  • IR Spectrum:
    Passes test
  • Substances reducing dichromate:
    Passes test ACS
  • Substances reducing KMnO4:
    Passes test
  • Alkalinity:
    Max. 0.0004 meq/g
  • Boiling point:
    117 - 119 °C
  • Colouration:
    Max. 10 APHA
  • Solidification point:
    16.2 - 16.6 °C
  • Substances coloured by H2SO4:
    Max. 150 APHA
  • Acetaldehyde:
    Max. 2 ppm
  • Acetic anhydride:
    Max. 100 ppm
  • Evaporation residue:
    Max. 5 ppm
  • Formate:
    Max. 0.05 %
  • Heavy metals (as Pb):
    Max. 0.5 ppm
  • Ignition residue (SO4):
    Max. 10 ppm
  • Water:
    Max. 0.25 %
  • Cl (Chloride):
    Max. 0.5 ppm
  • NO3 (Nitrate):
    Max. 2 ppm
  • PO4 (Phosphate):
    Max. 0.5 ppm
  • SO4 (Sulphate):
    Max. 1 ppm
  • Ag (Silver):
    Max. 0.5 ppm
  • Al (Aluminium):
    Max. 0.03 ppm
  • As (Arsenic):
    Max. 0.01 ppm
  • Ba (Barium):
    Max. 0.01 ppm
  • Be (Beryllium):
    Max. 0.01 ppm
  • Bi (Bismuth):
    Max. 0.02 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.02 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.08 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.2 ppm
  • Ge (Germanium):
    Max. 0.02 ppm
  • Hg (Mercury):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Li (Lithium):
    Max. 0.5 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Mo (Molybdenum):
    Max. 0.02 ppm
  • Na (Sodium):
    Max. 1 ppm
  • Ni (Nickel):
    Max. 0.1 ppm
  • Pb (Lead):
    Max. 0.02 ppm
  • Sb (Antimony):
    Max. 0.05 ppm
  • Sn (Tin):
    Max. 0.05 ppm
  • Sr (Strontium):
    Max. 0.01 ppm
  • Ti (Titanium):
    Max. 0.02 ppm
  • Tl (Thallium):
    Max. 0.02 ppm
  • V (Vanadium):
    Max. 0.01 ppm
  • Zn (Zinc):
    Max. 0.05 ppm
  • Zr (Zirconium):
    Max. 0.02 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together