
2,3-Hexanedione for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3848-24-6
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00009398
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–30 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:29 °C
EINECS:223-350-8
UN:1224
ADR:3,III
:M-Clarity™ quality level = MQ200
:114.14 g/mol
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-30 °C
- Molar Mass:114.15 g/mol
- Boiling Point:130 °C (1013 hPa)
- Vapor Pressure:13 hPa (20 °C)
- Flash Point:28 °C
- Explosion Limit:1.2 - 5.9 %(V)
- Density:0.94 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together