Order Entry
France
ContactUsLinkComponent
Chloroform, anhydrous (max. 0.005% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Health
Toxic
Chloroform, anhydrous (max. 0.005% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Catalog # 22709.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Toxic
Chloroform, anhydrous (max. 0.005% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Catalog # 22709.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.5 %
  • Colour value:
    Max. 10 APHA
  • Acidity:
    Max. 0.0005 %
  • Residue on evaporation:
    Max. 0.0005 %
  • Water (K.F.):
    Max. 0.0050 %
  • Dichloromethane:
    Max. 0.0100 %
  • Carbon tetrachloride:
    Max. 0.0100 %
  • Trichloroethene:
    Max. 0.0100 %
  • 1,1,2,2-Tetrachlorethane:
    Max. 0.0100 %
  • Formaldehyde-sulphuric colouration:
    Max. 60 APHA
  • Substances discoloured by H2SO4:
    Max. 35 APHA
  • Free chlorine:
    Max. 0.00003 %
  • Cl (Chloride):
    Max. 0.5 ppm
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.05 ppm

Specification Test Results

About this item

CAS Number:67-66-3
 :
Formula:CHCl₃
MDL Number:MFCD00000826
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
 :Stabilised with 2-methyl-2-butene (iso-amylene)
EINECS:200-663-8
REACH:01-2119486657-20
 :GHS08
 :GHS06