Orthoboric acid 10 g/l in aqueous solution with indicator (methyl red/bromocresl green mix)
Catalog # PRLS85766.360
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
About this item
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
:
:61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337